missing translation for 'onlineSavingsMsg'
Learn More

Isopropylcyclohexane 99.0+%, TCI America™

Catalog No. I0161500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0161500ML 500 mL
I016125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0161500ML Supplier TCI America Supplier No. I0161500ML
Only null left

Chemical Identifiers

CAS 696-29-7
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00001480
InChI Key GWESVXSMPKAFAS-UHFFFAOYSA-N
Synonym Hexahydrocumene
PubChem CID 12763
IUPAC Name (propan-2-yl)cyclohexane
SMILES CC(C)C1CCCCC1

Specifications

Color Colorless
Boiling Point 151°C
Quantity 500 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropylcyclohexane