missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropylboronic Acid (contains varying amounts of Anhydride), TCI America™
$218.10 - $218.10
Chemical Identifiers
| CAS | 80041-89-0 |
|---|---|
| Molecular Formula | C3H9BO2 |
| MDL Number | MFCD01319021 |
| InChI Key | QIPHSSYCQCBJAX-UHFFFAOYSA-N |
| PubChem CID | 2734750 |
Specifications
| 80041-89-0 | |
| White | |
| MFCD01319021 | |
| isopropylboronic acid, propan-2-yl boronic acid, 2-propaneboronic acid, prop-2-yl boronic acid, 1-methylethyl boronic acid, boronic acid, 1-methylethyl, isopropylboric acid, pubchem7987, isopropyl boronic acid, acmc-209piu | |
| B(C(C)C)(O)O | |
| 87.913 | |
| 87.91 | |
| Isopropylboronic Acid (contains varying amounts of Anhydride) |
| 115°C | |
| C3H9BO2 | |
| 5 g | |
| QIPHSSYCQCBJAX-UHFFFAOYSA-N | |
| propan-2-ylboronic acid | |
| 2734750 | |
| Crystalline Powder |
Safety and Handling
TSCA : No