missing translation for 'onlineSavingsMsg'
Learn More

Isopropylbenzene, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S2315

Catalog No. S-2315


Only null left

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Isopropylbenzene98-82-81000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Isopropylbenzene
98-82-8 , 67-56-1
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
C9H12
Certified Reference Material
CC(C)C1=CC=CC=C1
cumene
1 mL
CHEBI:34656
2mL Amber Prescored Glass Ampule
0.1,99.9
0.1,99.9
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
RWGFKTVRMDUZSP-UHFFFAOYSA-N
Methanol-P and T
120.195
7406

Chemical Identifiers

98-82-8
120.195
7406
cumene
C9H12
RWGFKTVRMDUZSP-UHFFFAOYSA-N
CHEBI:34656
CC(C)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Toxic if inhaled. Causes damage to organs.

DOTInformation : Nonhazardous

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only