missing translation for 'onlineSavingsMsg'
Learn More

Isopropylbenzene, Restek

Catalog No. 06719286
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719286 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719286 Supplier Chromatographic Supplier No. RK 30294

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Certified reference material for environmental analysis.

Chemical Identifiers

CAS 98-82-8 , 67-56-1
Molecular Formula C9H12
Molecular Weight (g/mol) 120.195
InChI Key RWGFKTVRMDUZSP-UHFFFAOYSA-N
PubChem CID 7406
ChEBI CHEBI:34656
IUPAC Name cumene
SMILES CC(C)C1=CC=CC=C1

Specifications

Concentration 2000 μg/mL
Solvent or Matrix Methanol-P and T
Industry Type Environmental Testing
Quantity 1 mL
Chemical Name or Material Isopropylbenzene