missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Sulfide 98.0+%, TCI America™
$69.31 - $111.66
Chemical Identifiers
| CAS | 625-80-9 |
|---|---|
| Molecular Formula | C6H14S |
| Molecular Weight (g/mol) | 118.24 |
| MDL Number | MFCD00008895 |
| InChI Key | XYWDPYKBIRQXQS-UHFFFAOYSA-N |
| Synonym | diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide |
| PubChem CID | 12264 |
| IUPAC Name | 2-(propan-2-ylsulfanyl)propane |
| SMILES | CC(C)SC(C)C |
Chemical Identifiers
| 625-80-9 | |
| 118.24 | |
| XYWDPYKBIRQXQS-UHFFFAOYSA-N | |
| 12264 | |
| CC(C)SC(C)C |
| C6H14S | |
| MFCD00008895 | |
| diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide | |
| 2-(propan-2-ylsulfanyl)propane |
Specifications
| 625-80-9 | |
| 121°C | |
| MFCD00008895 | |
| 1993 | |
| XYWDPYKBIRQXQS-UHFFFAOYSA-N | |
| 2-(propan-2-ylsulfanyl)propane | |
| 12264 | |
| ≥98.0% (GC) | |
| Isopropyl Sulfide |
| Yellow | |
| C6H14S | |
| 10 mL | |
| diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide | |
| CC(C)SC(C)C | |
| 118.24 | |
| 118.24 | |
| Liquid |
Safety and Handling
TSCA : Yes