missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Sulfide 98.0+%, TCI America™

Catalog No. I017510ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I017510ML 10 mL
I017525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I017510ML Supplier TCI America Supplier No. I017510ML

Chemical Identifiers

CAS 625-80-9
Molecular Formula C6H14S
Molecular Weight (g/mol) 118.24
MDL Number MFCD00008895
InChI Key XYWDPYKBIRQXQS-UHFFFAOYSA-N
Synonym diisopropyl sulfide, isopropyl sulfide, propane, 2,2'-thiobis, 2,4-dimethyl-3-thiapentane, isopropyl sulphide, diisopropyl sulphide, 2,2'-thiodipropane, 2-isopropylsulfanyl propane, 2-propan-2-ylsulfanyl propane, isopropylsulfide
PubChem CID 12264
IUPAC Name 2-(propan-2-ylsulfanyl)propane
SMILES CC(C)SC(C)C

Specifications

Color Yellow
Boiling Point 121°C
Quantity 10 mL
UN Number 1993
Formula Weight 118.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Sulfide