missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl salicylate, 99%, Thermo Scientific™

Isopropyl salicylate, 99%, ACROS Organics, Quantity: 250g, Packaging: Glass bottle, Boiling Point: 122.0 deg.C (18.0 mmHg), Colorless to Yellow, Molecular Weight: 180.203, Percent Purity: 99%, Assay Percent Range: 99%, CAS: 607-85-2, ChEBI: CHEBI:38703, Density: 1.0600g/mL, EINECS Number: 210-143-2

Identifiants chimiques

CAS 607-85-2
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00035703
InChI Key YEULQIJMIOWCHB-UHFFFAOYSA-N
Synonym isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
PubChem CID 11838
ChEBI CHEBI:38703
IUPAC Name propan-2-yl 2-hydroxybenzoate
SMILES CC(C)OC(=O)C1=CC=CC=C1O
voir les spécifications

Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Packaging Prix Quantity & Availability  
AC412891000
Afficher les documents Détails de la promotion
N/A N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Spécifications

Identifiants chimiques

607-85-2
180.2
YEULQIJMIOWCHB-UHFFFAOYSA-N
11838
propan-2-yl 2-hydroxybenzoate
C10H12O3
MFCD00035703
isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
CHEBI:38703
CC(C)OC(=O)C1=CC=CC=C1O

Spécifications

607-85-2
122.0°C (18.0 mmHg)
Authentic
C10H12O3
MFCD00035703
isopropyl salicylate, isopropyl 2-hydroxybenzoate, salicylic acid, isopropyl ester, iso-propyl salicylate, salicylic acid isopropyl ester, isopropyl o-hydroxybenzoate, benzoic acid, 2-hydroxy-, 1-methylethyl ester, 1-methylethyl 2-hydroxybenzoate, methylethyl 2-hydroxybenzoate, opropyl salicylate
CC(C)OC(=O)C1=CC=CC=C1O
180.2
CHEBI:38703
99%
1.0600g/mL
>93°C
99%
1.5090 to 1.5110
1.06
YEULQIJMIOWCHB-UHFFFAOYSA-N
propan-2-yl 2-hydroxybenzoate
11838
180.2
Isopropyl salicylate, 99%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 210-143-2

TSCA : TSCA

FDS
Documents

Documents

RUO – Research Use Only