Learn More
Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical
IS120, 67-63-0, C3H8O
Supplier:  Spectrum Chemical Mfg Cor IS12020LTCU
 View more versions of this product
Catalog No. 18603573
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
Isopropyl Rubbing Alcohol
70%,30%
20 L
0.872-0.883
UN1219
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
68 to 72%
67-63-0,7732-18-5
78.3°C
Cubitainer
C3H8O
70%
CC(C)O
60.10
USP
Chemical Identifiers
67-63-0
60.10
propan-2-ol
C3H8O
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CC(C)O
Product Suggestions
Product Suggestions