Learn More
Isopropyl Rubbing Alcohol, USP, 68-72%, Spectrum™ Chemical
IS120, 67-63-0, C3H8O
Supplier: Spectrum Chemical Mfg Cor IS1201LTGLCS6
Description
Spectrum™ Chemical Isopropyl Rubbing Alcohol, 70 Percent, USP , is a colorless and flammable chemical compound also known as Rubbing Alcohol. It is a cooling and soothing liquid for external applications to minimize the risk of infection and is not suitable for internal consumption. Spectrum Chemical USP products, including all USP alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
| Isopropyl Rubbing Alcohol | |
| 70%,30% | |
| 6 x 1 L | |
| 0.872-0.883 | |
| UN1219 | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| propan-2-ol | |
| 68 to 72% |
| 67-63-0,7732-18-5 | |
| 78.3°C | |
| Amber Glass Bottle | |
| C3H8O | |
| 70% | |
| CC(C)O | |
| 60.10 | |
| USP |
Chemical Identifiers
| 67-63-0 | |
| 60.10 | |
| propan-2-ol |
| C3H8O | |
| KFZMGEQAYNKOFK-UHFFFAOYSA-N | |
| CC(C)O |