missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Propionate 99.0+%, TCI America™

Catalog No. p-7134647
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P050725ML 25 mL
P0507500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P050725ML Supplier TCI America Supplier No. P050725ML
Only null left

Chemical Identifiers

CAS 637-78-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00051553
InChI Key IJMWOMHMDSDKGK-UHFFFAOYSA-N
Synonym isopropyl propionate, propanoic acid, 1-methylethyl ester, isopropyl propanoate, propionic acid isopropyl ester, propionic acid, isopropyl ester, 1-methylethyl propanoate, unii-ngb0ak08t9, iso-propyl propanoate, iso-propyl n-propionate, ngb0ak08t9
PubChem CID 12508
IUPAC Name propan-2-yl propanoate
SMILES CCC(=O)OC(C)C

Specifications

Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 2409
Formula Weight 116.16
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Propionate
EINECSNumber (2)-0774
TSCA Yes