missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Propionate 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 108°C |
| Quantity | 500 mL |
| UN Number | 2409 |
| Formula Weight | 116.16 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isopropyl Propionate |
Safety and Handling
| EINECSNumber | (2)-0774 |
| TSCA | Yes |