Learn More
Isopropyl palmitate, tech. 85%
CAS: 142-91-6 | C19H38O2 | 298.51 g/mol
Supplier: Thermo Scientific Chemicals L04276AU
Description
Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| Isopropyl palmitate | |
| 11°C to 13°C | |
| 160°C (2 mmHg) | |
| C19H38O2 | |
| MFCD00008993 | |
| 1786567 | |
| Not miscible or difficult to mix with water. | |
| CCCCCCCCCCCCCCCC(=O)OC(C)C | |
| 298.51 | |
| CHEBI:84262 | |
| 85% |
| 142-91-6 | |
| 0.852 | |
| 183°C (361°F) | |
| 1.438 | |
| 1000 mL | |
| isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit | |
| XUGNVMKQXJXZCD-UHFFFAOYSA-N | |
| propan-2-yl hexadecanoate | |
| 8907 | |
| 298.51 | |
| Technical |
Chemical Identifiers
| 142-91-6 | |
| 298.51 | |
| XUGNVMKQXJXZCD-UHFFFAOYSA-N | |
| 8907 | |
| propan-2-yl hexadecanoate |
| C19H38O2 | |
| MFCD00008993 | |
| isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit | |
| CHEBI:84262 | |
| CCCCCCCCCCCCCCCC(=O)OC(C)C |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 205-571-1
RTECSNumber : RT4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only