missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl palmitate, tech. 85%

Catalog No. AAL04276AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL04276AK 250 mL
AAL04276AU 1000 mL
AAL042760F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL04276AK Supplier Thermo Scientific Chemicals Supplier No. L04276AK

CAS: 142-91-6 | C19H38O2 | 298.51 g/mol

Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl palmitate is used to formulate and evaluate the suitability of pluronic lecithin organogels containing flurbiprofen for topical application and to characterize microemulsion systems of isopropyl palmitate, water and 2:1 Brij 97, and 1-butanol by different experimental techniques.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00008993
InChI Key XUGNVMKQXJXZCD-UHFFFAOYSA-N
Synonym isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
PubChem CID 8907
ChEBI CHEBI:84262
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Melting Point 11°C to 13°C
Density 0.852
Boiling Point 160°C (2 mmHg)
Flash Point 183°C (361°F)
Refractive Index 1.438
Quantity 250 mL
Beilstein 1786567
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 298.51
Percent Purity 85%
Grade Technical
Chemical Name or Material Isopropyl palmitate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 205-571-1
RTECSNumber RT4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only