missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate 97.0+%, TCI America™

Catalog No. P000525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P000525ML 25 mL
P0005500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P000525ML Supplier TCI America Supplier No. P000525ML

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00008993
InChI Key XUGNVMKQXJXZCD-UHFFFAOYSA-N
Synonym isopropyl palmitate, isopropyl hexadecanoate, isopalm, deltyl, isopal, propal, deltyl prime, tegester isopalm, ja-fa ippkessco, sinnoester pit
PubChem CID 8907
ChEBI CHEBI:84262
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Melting Point 13°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 298.51
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Palmitate
EINECSNumber (2)-0798
RTECSNumber RT4900000
TSCA Yes