missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Palmitate, NF, 90%, Spectrum™ Chemical

Catalog No. p-7157641
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Catalog No. Quantity Packaging
18603558 500 mL Amber Glass Bottle
18603559 6 x 500 mL Amber Glass Bottle
18603560 4 L Amber Glass Bottle
18603561 4 x 4 L Amber Glass Bottle
18603562 20 L Steel Can
18603563 200 L Drum
6 options

Catalog No. 18603558

Supplier: Spectrum Chemical Mfg Cor IS160500MLGL

Only null left

IS160, 142-91-6, C19H38O2

Spectrum™ Chemical Isopropyl Palmitate, NF is derived from palm oil and is used as a thickening agent and antistatic agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 142-91-6
Molecular Formula C19H38O2
Molecular Weight (g/mol) 298.51
MDL Number MFCD00008993
InChI Key XUGNVMKQXJXZCD-UHFFFAOYSA-N
IUPAC Name propan-2-yl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Refractive Index 1.435-1.438
Quantity 500 mL
Percent Purity 90%
Grade NF