missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Isopropyl Palmitate NF
Description
Isopropyl Palmitate NF is a colorless, mobile liquid having a very slight odor.
Specifications
Specifications
| Chemical Name or Material | Isopropyl Palmitate NF |
| Packaging | Bottle, Amber Glass, Boston Round, 500mL, 38-439 |
| CAS | 142-91-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 298.5072 |
| Quantity | 500 mL |
| Percent Purity | ≥90.0 % |
| Grade | NF |