missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Myristate, NF, 90%, Spectrum™ Chemical

IS145, 110-27-0, C17H34O2

Supplier:  Spectrum Chemical Mfg Cor IS14520LTMT

 View more versions of this product

Catalog No. 18603556


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Isopropyl Myristate, NF is an emollient ester of low viscosity used to reduce the greasiness of emulsions. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

110-27-0
0.1%
C17H34O2
20 L
CCCCCCCCCCCCCC(=O)OC(C)C
270.46
NF
100%
Steel Can
1.432-1.436
AXISYYRBXTVTFY-UHFFFAOYSA-N
propan-2-yl tetradecanoate
90%

Chemical Identifiers

110-27-0
270.46
propan-2-yl tetradecanoate
C17H34O2
AXISYYRBXTVTFY-UHFFFAOYSA-N
CCCCCCCCCCCCCC(=O)OC(C)C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions