missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Myristate, NF, 90%, Spectrum™ Chemical

Catalog No. p-7157640
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603552 500 mL Amber Glass Bottle
18603553 6 x 500 mL Amber Glass Bottle
18603554 4 L Amber Glass Bottle
18603555 4 x 4 L Amber Glass Bottle
18603556 20 L Steel Can
18603557 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18603552 Supplier Spectrum Chemical Mfg Cor Supplier No. IS145500MLGL
Only null left

IS145, 110-27-0, C17H34O2

Spectrum™ Chemical Isopropyl Myristate, NF is an emollient ester of low viscosity used to reduce the greasiness of emulsions. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.46
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Refractive Index 1.432-1.436
Quantity 500 mL
Percent Purity 90%
Grade NF