missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Isopropyl Myristate NF
Description
Isopropyl Myristate NF is a clear, practically colorless, oily liquid. Its practically odorless and congeals at about 5°C.
Alternative name(s): IPM, isopropyl tetradecanoate,
Specifications
Specifications
| Chemical Name or Material | Isopropyl Myristate NF |
| Packaging | Bottle, 500mL, Amber, PET, Boston Round, 28-400 |
| CAS | 110-27-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 270.4536 |
| Quantity | 500 mL |
| Percent Purity | ≥90.0 % |
| Grade | NF |