Learn More
Isopropyl myristate, 96%
CAS: 110-27-0 | C17H34O2 | 270.45 g/mol
Supplier: Thermo Scientific Chemicals 412850010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Isopropyl myristate | |
| 110-27-0 | |
| 0.8500g/mL | |
| >110°C | |
| 95% min. (GC) | |
| C17H34O2 | |
| CH3(CH2)12COOCH(CH3)2 | |
| 1 kg | |
| 15, 5261 | |
| AXISYYRBXTVTFY-UHFFFAOYSA-N | |
| propan-2-yl tetradecanoate | |
| 8042 | |
| 96% |
| 96% | |
| 0.5mg KOH/g max. | |
| 167.0°C (9.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4340 to 1.4360 | |
| MFCD00008982 | |
| 0.85 | |
| isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
| CCCCCCCCCCCCCC(=O)OC(C)C | |
| 270.45 | |
| 270.45 | |
| Liquid |
Chemical Identifiers
| 110-27-0 | |
| 270.45 | |
| AXISYYRBXTVTFY-UHFFFAOYSA-N | |
| 8042 | |
| CCCCCCCCCCCCCC(=O)OC(C)C |
| C17H34O2 | |
| MFCD00008982 | |
| isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester | |
| propan-2-yl tetradecanoate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 203-751-4
RTECSNumber : XB8600000
TSCA : TSCA
RUO – Research Use Only