missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl myristate, 96%

Catalog No. p-152066
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412850250 25 g Glass bottle
AC412851000 100 g Glass bottle
AC412850010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC412850250 Supplier Thermo Scientific Chemicals Supplier No. 412850250
Only null left

CAS: 110-27-0 | C17H34O2 | 270.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-27-0
Molecular Formula C17H34O2
Molecular Weight (g/mol) 270.45
MDL Number MFCD00008982
InChI Key AXISYYRBXTVTFY-UHFFFAOYSA-N
Synonym isopropyl myristate, isopropyl tetradecanoate, estergel, bisomel, isomyst, promyr, tetradecanoic acid, 1-methylethyl ester, deltyl extra, kesscomir, tegester
PubChem CID 8042
IUPAC Name propan-2-yl tetradecanoate
SMILES CCCCCCCCCCCCCC(=O)OC(C)C

Specifications

Acidity 0.5mg KOH/g max.
Density 0.8500g/mL
Boiling Point 167.0°C (9.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.4360
Linear Formula CH3(CH2)12COOCH(CH3)2
Quantity 25 g
Specific Gravity 0.85
Merck Index 15, 5261
Formula Weight 270.45
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Isopropyl myristate, 96%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-751-4
RTECSNumber XB8600000
TSCA TSCA

RUO – Research Use Only