missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Methyl Sulfone 97.0+%, TCI America™

Catalog No. I04845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04845G 5 g
I048425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04845G Supplier TCI America Supplier No. I04845G
Only null left

Chemical Identifiers

CAS 4853-74-1
Molecular Formula C4H10O2S
Molecular Weight (g/mol) 122.182
MDL Number MFCD00050415
InChI Key VTWYQAQIXXAXOR-UHFFFAOYSA-N
PubChem CID 138372
IUPAC Name 2-methylsulfonylpropane
SMILES CC(C)S(=O)(=O)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 122.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Methyl Sulfone