accessibility menu, dialog, popup

UserName

CAS RN 4853-74-1

H3CCH3SCH3OO

CAS RN 4853-74-1

IUPAC Name: 2-methanesulfonylpropane
Synonyms: 2-methanesulfonylpropane
Molecular Weight (g/mol): 122.18
Molecular Formula: C4H10O2S
InChi Key: VTWYQAQIXXAXOR-UHFFFAOYSA-N
SMILES: CC(C)S(C)(=O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results