missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Linoleate 60.0+%, TCI America™

Catalog No. p-7131837
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L013925ML 25 mL
L0139500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L013925ML Supplier TCI America Supplier No. L013925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22882-95-7
Molecular Formula C21H38O2
Molecular Weight (g/mol) 322.533
MDL Number MFCD00053809
InChI Key XEIOPEQGDSYOIH-MVKOLZDDSA-N
Synonym Linoleic Acid Isopropyl Ester
PubChem CID 13647303
IUPAC Name propan-2-yl (9E,12E)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC(C)C

Specifications

Color Yellow
Boiling Point 179°C
Quantity 25 mL
Formula Weight 322.53
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Linoleate
RTECSNumber RG0180210
TSCA Yes