accessibility menu, dialog, popup

UserName

CH3OOCH3CH3(Z)(Z)

CAS RN 22882-95-7

IUPAC Name: propan-2-yl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: isopropyl linoleate
Molecular Weight (g/mol): 322.53
Molecular Formula: C21H38O2
InChi Key: XEIOPEQGDSYOIH-MURFETPASA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results