missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I05205ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05205ML 5 mL
I052025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05205ML Supplier TCI America Supplier No. I05205ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2253-73-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00037830
InChI Key VHBFEIBMZHEWSX-UHFFFAOYSA-N
Synonym isopropyl isothiocyanate, isothiocyanic acid isopropyl ester, isopropylisothiocyanate, propane, 2-isothiocyanato, unii-46561pkp9t, isopropyl mustard oil, 2-isothiocyanato-propane, propan-2-isothiocyanate, ipncs, isopropyl isothiocya
PubChem CID 75263
IUPAC Name 2-isothiocyanatopropane
SMILES CC(C)N=C=S

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 mL
UN Number 1992
Formula Weight 101.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Isothiocyanate