missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl isothiocyanate, 97%

Catalog No. p-7081605
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1164106 5 g
AAA1164114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1164106 Supplier Thermo Scientific Chemicals Supplier No. A1164106
Only null left

CAS: 2253-73-8 | C4H7NS | 101.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Isopropyl isothiocyanate is used as is an antimicrobial agent. It is also used as medicinal intermediate.

Solubility
Miscible with methanol. Hydrolyzes in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with alcohols and amines.

Chemical Identifiers

CAS 2253-73-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00037830
InChI Key VHBFEIBMZHEWSX-UHFFFAOYSA-N
Synonym isopropyl isothiocyanate, isothiocyanic acid isopropyl ester, isopropylisothiocyanate, propane, 2-isothiocyanato, unii-46561pkp9t, isopropyl mustard oil, 2-isothiocyanato-propane, propan-2-isothiocyanate, ipncs, isopropyl isothiocya
PubChem CID 75263
IUPAC Name 2-isothiocyanatopropane
SMILES CC(C)N=C=S

Specifications

Density 0.948
Boiling Point 137°C to 138°C
Flash Point 40°C (104°F)
Odor Pungent
Refractive Index 1.493
Quantity 5 g
UN Number UN2924
Beilstein 1739163
Sensitivity Moisture sensitive
Solubility Information Miscible with methanol. Hydrolyzes in water.
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Isopropyl isothiocyanate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 218-851-3
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only