missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Hexanoate 98.0+%, TCI America™
Supplier: TCI America C084125ML
Specifications
| Isopropyl Hexanoate | |
| Colorless | |
| C9H18O2 | |
| 25 mL | |
| Hexanoic Acid Isopropyl Ester | |
| CCCCCC(=O)OC(C)C | |
| 158.241 | |
| 158.24 | |
| Liquid |
| 2311-46-8 | |
| 168°C | |
| MFCD00059436 | |
| 3272 | |
| JSHDAORXSNJOBA-UHFFFAOYSA-N | |
| propan-2-yl hexanoate | |
| 16832 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2311-46-8 | |
| 158.241 | |
| JSHDAORXSNJOBA-UHFFFAOYSA-N | |
| 16832 | |
| CCCCCC(=O)OC(C)C |
| C9H18O2 | |
| MFCD00059436 | |
| Hexanoic Acid Isopropyl Ester | |
| propan-2-yl hexanoate |
Safety and Handling
EINECSNumber : (2)-0782
TSCA : No