accessibility menu, dialog, popup

UserName

CAS RN 2311-46-8

H3COOCH3CH3

CAS RN 2311-46-8

IUPAC Name: propan-2-yl hexanoate
Synonyms: isopropyl hexanoate
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: JSHDAORXSNJOBA-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results