missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl glycolate, 96%

Catalog No. p-7033258
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5033403 1 g
AAH5033406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5033403 Supplier Thermo Scientific Chemicals Supplier No. H5033403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 623-61-0 | C5H10O3 | 118.132 g/mol

Isopropyl glycolate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl glycolate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 623-61-0
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00046747
InChI Key AZKIQQBSVTWCGY-UHFFFAOYSA-N
Synonym isopropyl glycolate, isopropyl 2-hydroxyacetate, isopropyl hydroxyacetate, 2-hydroxyacetic acid isopropyl ester, acetic acid, hydroxy-, 1-methylethyl ester, isopropylglycolate, acmc-209x9i, glycolic acid isopropyl ester, ksc614s5p, hydroxy-acetic acid isopropyl ester
PubChem CID 69338
IUPAC Name propan-2-yl 2-hydroxyacetate
SMILES CC(C)OC(=O)CO

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 118.13
Percent Purity 96%
Chemical Name or Material Isopropyl glycolate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-804-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only