CAS RN 623-61-0
IUPAC Name:
propan-2-yl 2-hydroxyacetate
Synonyms:
isopropyl 2-hydroxyacetate
Molecular Weight (g/mol):
118.13
Molecular Formula:
C5H10O3
InChi Key:
AZKIQQBSVTWCGY-UHFFFAOYSA-N
SMILES:
CC(C)OC(=O)CO
1
–
1
of
1
results
1
Promotions Available
Isopropyl glycolate, 96%
Isopropyl glycolate, 96%
CAS: 623-61-0 Molecular Formula: C5H10O3 Molecular Weight (g/mol): 118.132 MDL Number: MFCD00046747 InChI Key: AZKIQQBSVTWCGY-UHFFFAOYSA-N Synonym: isopropyl glycolate,isopropyl 2-hydroxyacetate,isopropyl hydroxyacetate,2-hydroxyacetic acid isopropyl ester,acetic acid, hydroxy-, 1-methylethyl ester,isopropylglycolate,acmc-209x9i,glycolic acid isopropyl ester,ksc614s5p,hydroxy-acetic acid isopropyl ester PubChem CID: 69338 IUPAC Name: propan-2-yl 2-hydroxyacetate SMILES: CC(C)OC(=O)CO