missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Formate 97.0+%, TCI America™

Catalog No. p-7129468
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
F005625ML 25 mL
F0056500ML 500 mL
2 options

Catalog No. F005625ML

Supplier: TCI America F005625ML

Only null left

Chemical Identifiers

CAS 625-55-8
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00014127
InChI Key RMOUBSOVHSONPZ-UHFFFAOYSA-N
Synonym isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2
PubChem CID 12257
IUPAC Name propan-2-yl formate
SMILES CC(C)OC=O

Specifications

Color Colorless
Boiling Point 68°C
Quantity 25 mL
UN Number 1281
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Formate
EINECSNumber (2)-0684
RTECSNumber LQ8750000
TSCA Yes