accessibility menu, dialog, popup

UserName

CH3CH3OO

CAS RN 625-55-8

IUPAC Name: propan-2-yl formate
Synonyms: isopropyl formate
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: RMOUBSOVHSONPZ-UHFFFAOYSA-N
SMILES: CC(C)OC=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)