missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Formate 97.0+%, TCI America™

Catalog No. F005625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F005625ML 25 mL
F0056500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F005625ML Supplier TCI America Supplier No. F005625ML

Chemical Identifiers

CAS 625-55-8
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00014127
InChI Key RMOUBSOVHSONPZ-UHFFFAOYSA-N
Synonym isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2
PubChem CID 12257
IUPAC Name propan-2-yl formate
SMILES CC(C)OC=O

Specifications

Color Colorless
Boiling Point 68°C
Quantity 25 mL
UN Number 1281
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Formate
EINECSNumber (2)-0684
RTECSNumber LQ8750000
TSCA Yes