missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl formate, 98%

Catalog No. aaa1722218
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1722218 50 g
AAA1722230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1722218 Supplier Thermo Scientific Chemicals Supplier No. A1722218
Only null left

CAS: 625-55-8 | C4H8O2 | 88.11 g/mol

Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.

Solubility
slightly soluble in water, completely miscible with alcohol, ether, and most organic solvents.

Notes
d^2^0 0.901. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 625-55-8
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00014127
InChI Key RMOUBSOVHSONPZ-UHFFFAOYSA-N
Synonym isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2
PubChem CID 12257
IUPAC Name propan-2-yl formate
SMILES CC(C)OC=O

Specifications

Melting Point -93°C
Density 0.88
Boiling Point 68°C to 71°C
Flash Point −6°C (21°F)
Refractive Index 1.3685
Linear Formula HCO2CH(CH3)2
Quantity 50 g
UN Number UN1281
Beilstein 1735844
Solubility Information slightly soluble in water,completely miscible with alcohol,ether,and most organic solvents.
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material Isopropyl formate
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H319-H335-H336
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYL FORMATES
EINECSNumber 210-901-2
RTECSNumber LQ8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only