missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Cyclohexanecarboxylate 98.0+%, TCI America™
$96.94 - $1043.87
Chemical Identifiers
| CAS | 6553-80-6 |
|---|---|
| Molecular Formula | C10H18O2 |
| Molecular Weight (g/mol) | 170.252 |
| MDL Number | MFCD00058949 |
| InChI Key | PMQZNGSMBAGPRU-UHFFFAOYSA-N |
| Synonym | Cyclohexanecarboxylic Acid Isopropyl Ester |
| PubChem CID | 81039 |
| IUPAC Name | propan-2-yl cyclohexanecarboxylate |
| SMILES | CC(C)OC(=O)C1CCCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C047225ML
|
TCI America
C047225ML |
25 mL |
Each for $96.94
|
|
|||||
|
C0472500ML
|
TCI America
C0472500ML |
500 mL |
Each for $1,043.87
|
|
|||||
Chemical Identifiers
| 6553-80-6 | |
| 170.252 | |
| PMQZNGSMBAGPRU-UHFFFAOYSA-N | |
| 81039 | |
| CC(C)OC(=O)C1CCCCC1 |
| C10H18O2 | |
| MFCD00058949 | |
| Cyclohexanecarboxylic Acid Isopropyl Ester | |
| propan-2-yl cyclohexanecarboxylate |
Specifications
| 6553-80-6 | |
| 131°C | |
| MFCD00058949 | |
| Cyclohexanecarboxylic Acid Isopropyl Ester | |
| CC(C)OC(=O)C1CCCCC1 | |
| 170.252 | |
| 170.25 | |
| Liquid |
| Colorless | |
| C10H18O2 | |
| 25 mL | |
| PMQZNGSMBAGPRU-UHFFFAOYSA-N | |
| propan-2-yl cyclohexanecarboxylate | |
| 81039 | |
| ≥98.0% (GC) | |
| Isopropyl Cyclohexanecarboxylate |
Safety and Handling
TSCA : No