accessibility menu, dialog, popup

UserName

H3CCH3OO

CAS RN 6553-80-6

IUPAC Name: propan-2-yl cyclohexanecarboxylate
Synonyms: isopropyl cyclohexanecarboxylate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: PMQZNGSMBAGPRU-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1CCCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results