missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Cyclohexanecarboxylate 98.0+%, TCI America™

Catalog No. C0472500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0472500ML 500 mL
C047225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0472500ML Supplier TCI America Supplier No. C0472500ML
Only null left

Chemical Identifiers

CAS 6553-80-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00058949
InChI Key PMQZNGSMBAGPRU-UHFFFAOYSA-N
Synonym Cyclohexanecarboxylic Acid Isopropyl Ester
PubChem CID 81039
IUPAC Name propan-2-yl cyclohexanecarboxylate
SMILES CC(C)OC(=O)C1CCCCC1

Specifications

Color Colorless
Boiling Point 131°C
Quantity 500 mL
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Cyclohexanecarboxylate