missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Cyanoacetate 97.0+%, TCI America™

Catalog No. C100125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C100125ML 25 mL
C1001500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C100125ML Supplier TCI America Supplier No. C100125ML

Chemical Identifiers

CAS 13361-30-3
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00019842
InChI Key BESQLCCRQYTQQI-UHFFFAOYSA-N
Synonym isopropyl cyanoacetate, isopropyl 2-cyanoacetate, cyanoacetic acid isopropyl ester, acetic acid, cyano-, isopropyl ester, unii-0v3mgi9xma, 0v3mgi9xma, acetic acid, cyano-, 1-methylethyl ester, propan-2-yl cyanoacetate, prop-2-yl cyanoacetate, acmc-1bsph
PubChem CID 25917
IUPAC Name propan-2-yl 2-cyanoacetate
SMILES CC(C)OC(=O)CC#N

Specifications

Color Colorless
Boiling Point 206°C
Quantity 25 mL
Formula Weight 127.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Cyanoacetate
EINECSNumber (2)-1549
RTECSNumber AG4270000
TSCA No