accessibility menu, dialog, popup

UserName

CAS RN 13361-30-3

IUPAC Name: propan-2-yl 2-cyanoacetate
Synonyms: isopropyl 2-cyanoacetate
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: BESQLCCRQYTQQI-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)CC#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)