missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl cyanoacetate, 97%

Catalog No. p-7081759
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1188714 25 g
AAA1188722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1188714 Supplier Thermo Scientific Chemicals Supplier No. A1188714
Only null left

CAS: 13361-30-3 | C6H9NO2 | 127.143 g/mol

Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing KnÖevenagel reactions with aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isopropyl cyanoacetate was used in the synthesis of 3-substituted coumarins by undergoing Knöevenagel reactions with aldehydes and ketones.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 13361-30-3
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00019842
InChI Key BESQLCCRQYTQQI-UHFFFAOYSA-N
Synonym isopropyl cyanoacetate, isopropyl 2-cyanoacetate, cyanoacetic acid isopropyl ester, acetic acid, cyano-, isopropyl ester, unii-0v3mgi9xma, 0v3mgi9xma, acetic acid, cyano-, 1-methylethyl ester, propan-2-yl cyanoacetate, prop-2-yl cyanoacetate, acmc-1bsph
PubChem CID 25917
IUPAC Name propan-2-yl 2-cyanoacetate
SMILES CC(C)OC(=O)CC#N

Specifications

Density 1.013
Boiling Point 207°C
Flash Point 92°C (198°F)
Refractive Index 1.415
Quantity 25 g
Beilstein 1756014
Solubility Information Slightly soluble in water.
Formula Weight 127.14
Percent Purity 97%
Chemical Name or Material Isopropyl cyanoacetate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 236-421-3
RTECSNumber AG4270000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only