missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Chloroacetate 98.0+%, TCI America™

Catalog No. p-7124700
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
C094825ML 25 mL
C0948500ML 500 mL
2 options

Catalog No. C094825ML

Supplier: TCI America C094825ML

Only null left

Chemical Identifiers

CAS 105-48-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00040410
InChI Key VODRWDBLLGYRJT-UHFFFAOYSA-N
Synonym isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
PubChem CID 7759
IUPAC Name propan-2-yl 2-chloroacetate
SMILES CC(C)OC(=O)CCl

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
UN Number 2947
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Chloroacetate
EINECSNumber (2)-1149
TSCA Yes