missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Chloroacetate 98.0+%, TCI America™

Catalog No. c094825ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C094825ML 25 mL
C0948500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C094825ML Supplier TCI America Supplier No. C094825ML
Only null left

Chemical Identifiers

CAS 105-48-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00040410
InChI Key VODRWDBLLGYRJT-UHFFFAOYSA-N
Synonym isopropyl chloroacetate, chloroacetic acid isopropyl ester, acetic acid, chloro-, 1-methylethyl ester, iso-propyl chloroacetate, clch2c o och ch3 2, ccris 7748, acetic acid, chloro-, isopropyl ester, acetic acid, 2-chloro-, 1-methylethyl ester, chloroacetic acid, 1-methyl ester, monochloroacetic acid isopropyl ester
PubChem CID 7759
IUPAC Name propan-2-yl 2-chloroacetate
SMILES CC(C)OC(=O)CCl

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
UN Number 2947
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Isopropyl Chloroacetate
EINECSNumber (2)-1149
TSCA Yes