missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Butyrate 99.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 130°C |
| Quantity | 25 mL |
| UN Number | 2405 |
| Formula Weight | 130.19 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Isopropyl Butyrate |
Safety and Handling
| EINECSNumber | (2)-0779 |
| TSCA | No |