CAS RN 638-11-9
IUPAC Name:
propan-2-yl butanoate
Synonyms:
isopropyl butyrate
Molecular Weight (g/mol):
130.19
Molecular Formula:
C7H14O2
InChi Key:
FFOPEPMHKILNIT-UHFFFAOYSA-N
SMILES:
CCCC(=O)OC(C)C
1
–
1
of
1
results
1
Isopropyl Butyrate 99.0+%, TCI America™
CAS: 638-11-9 Molecular Formula: C7H14O2 Molecular Weight (g/mol): 130.187 MDL Number: MFCD00009390 InChI Key: FFOPEPMHKILNIT-UHFFFAOYSA-N Synonym: Butyric Acid Isopropyl Ester PubChem CID: 61184 IUPAC Name: propan-2-yl butanoate SMILES: CCCC(=O)OC(C)C