missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals
$1,132.84 - $3,221.95
Chemical Identifiers
| CAS | 367-93-1 |
|---|---|
| Molecular Formula | C9H18O5S |
| Molecular Weight (g/mol) | 238.298 |
| MDL Number | MFCD00063273 |
| InChI Key | BPHPUYQFMNQIOC-NXRLNHOXSA-N |
| Synonym | iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside |
| PubChem CID | 656894 |
| ChEBI | CHEBI:61448 |
| IUPAC Name | (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol |
| SMILES | CC(C)SC1C(C(C(C(O1)CO)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AAJ1788614
|
N/A | 25 g | Plastic bottle |
Each for $1,132.84
|
|
||||
|
AAJ1788622
|
N/A | 100 g | Plastic bottle |
Each for $3,221.95
|
|
||||
Description
- Induces the lac operon in E. coli by binding and inactivating the lac repressor.
- Selection for lac Y mutants.
- Selection for recombinant plasmids containing β-galactosidase fusion proteins.
- Induces the cellular content of lactose permease.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Induces lac operon in E. coli by binding and inactivating lac repressor
- Selection for lac Y mutants
- Selection for recombinant plasmids containing beta-galactosidase fusion proteins
- Induces cellular content of lactose permease
Chemical Identifiers
| 367-93-1 | |
| 238.298 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| 656894 | |
| (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol |
| C9H18O5S | |
| MFCD00063273 | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CHEBI:61448 | |
| CC(C)SC1C(C(C(C(O1)CO)O)O)O |
Specifications
| 367-93-1 | |
| C9H18O5S | |
| 4631 | |
| BPHPUYQFMNQIOC-NXRLNHOXSA-N | |
| (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol | |
| 656894 | |
| 238.3 | |
| Ultrapure | |
| Isopropyl-beta-D-thiogalactopyranoside (IPTG) |
| 109°C to 115°C | |
| MFCD00063273 | |
| iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside | |
| CC(C)SC1C(C(C(C(O1)CO)O)O)O | |
| 238.298 | |
| CHEBI:61448 | |
| ≥99% | |
| Powder |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-703-0
TSCA : Yes
Recommended Storage : Store at -20°C
RUO – Research Use Only