missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl-beta-D-thiogalactopyranoside (IPTG), Dioxane free, >99%, Ultrapure, Thermo Scientific Chemicals

Catalog No. AAJ1788614
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1788614 25 g Plastic bottle
AAJ1788622 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1788614 Supplier Thermo Fisher Scientific Supplier No. J1788614

  • Induces the lac operon in E. coli by binding and inactivating the lac repressor.
  • Selection for lac Y mutants.
  • Selection for recombinant plasmids containing β-galactosidase fusion proteins.
  • Induces the cellular content of lactose permease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Induces lac operon in E. coli by binding and inactivating lac repressor
  • Selection for lac Y mutants
  • Selection for recombinant plasmids containing beta-galactosidase fusion proteins
  • Induces cellular content of lactose permea

Chemical Identifiers

CAS 367-93-1
Molecular Formula C9H18O5S
Molecular Weight (g/mol) 238.298
MDL Number MFCD00063273
InChI Key BPHPUYQFMNQIOC-NXRLNHOXSA-N
Synonym iptg, isopropyl-beta-d-thiogalactopyranoside, isopropyl-beta-d-thiogalactoside, isopropyl thiogalactoside, isopropyl beta-d-thiogalactopyranoside, isopropyl beta-d-thiogalactoside, isopropyl beta-d-1-thiogalactopyranoside, unii-x73vv2246b, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-isopropylthio tetrahydro-2h-pyran-3,4,5-triol, isopropyl-1-beta-d-thiogalactoside
PubChem CID 656894
ChEBI CHEBI:61448
IUPAC Name (2 R,3 R,4 S,5 R,6 S)-2-(hydroxymethyl)-6-propan-2-ylsulfanyloxane-3,4,5-triol
SMILES CC(C)SC1C(C(C(C(O1)CO)O)O)O

Specifications

Melting Point 109°C to 115°C
Packaging Plastic bottle
Quantity 25 g
Beilstein 4631
Formula Weight 238.3
Percent Purity ≥99%
Grade Ultrapure
Physical Form Powder
Chemical Name or Material Isopropyl-beta-D-thiogalactopyranoside (IPTG)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-703-0
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only