missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol 99.5+%, TCI America™

Catalog No. p-7131247
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I016325ML 25 mL
I0163500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I016325ML Supplier TCI America Supplier No. I016325ML
Only null left

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym Isopropanol, 2-Propanol, IPA
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Color Colorless
Quantity 25 mL
Physical Form Clear Liquid at 20°C
UN Number 1219
Boiling Point 82°C
Formula Weight 60.10
Percent Purity ≥99.5% (GC)
Chemical Name or Material Isopropyl Alcohol
EINECSNumber (2)-0207
RTECSNumber NT8050000
TSCA Yes