missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, Reagent, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7157610
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Bottle
Steel Can
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603551 125 mL Amber Glass Bottle
18603541 500 mL Amber Glass Bottle
18603543 1 L Amber Glass Bottle
18603542 6 x 500 mL Amber Glass Bottle
18603544 4 L Amber Glass Bottle
18603545 4 L Poly Bottle
18603546 6 x 1 L Amber Glass Bottle
18603547 4 x 4 L Amber Glass Bottle
18603548 4 x 4 L Poly Bottle
18603549 20 L Steel Can
18603550 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 18603551 Supplier Spectrum Chemical Mfg Cor Supplier No. I1048125ML
Only null left

I1048, 67-63-0, C3H8O

Spectrum™ Chemical Isopropyl Alcohol, Reagent, ACS is a compound which is colorless, flammable and has a very strong odor. Isopropyl alcohol has many uses, including as an analytical solvent reagent in the laboratory. Other chemical applications include use as a chemical synthesis intermediate, disinfectant, fuel additive and as a biological specimen preservative.

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100%
Quantity 125 mL
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
UN Number UN1219
Water 0.2%
Boiling Point 78.3°C
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material Isopropyl Alcohol