missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Alcohol, OmniSolv, MilliporeSigma™

Catalog No. p-217394
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX18346 1 L Glass Bottle
MPX18341 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MPX18346 Supplier Technidata Supplier No. PX18346
Only null left

Ideal for Spectrophotometry

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3CHOHCH3
Formula Weight 60.1
Percent Purity ≥99.9%
Grade OmniSolv
Chemical Name or Material Isopropyl Alcohol
Recommended Storage Room Temperature